Polyvinyl alcohol, EMPROVE® ESSENTIAL Ph. Eur., USP, JPE

Supplier: MERCK PRODUCTION CHEMICALS

Synonyms: Mowiol®PVOHPVA

MERP1.41439.1000EA 0 EUR
MERP1.41439.1000
Polyvinyl alcohol, EMPROVE® ESSENTIAL Ph. Eur., USP, JPE
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR