MXP high pressure injection valve, Rheodyne®, MX Series II™
Supplier: IDEX Health & Science
554-3290EA 0 EUR
RHEOMXP7920-000
MXP high pressure injection valve, Rheodyne®, MX Series II™
This high pressure actuated valve is designed to be added to an existing instrument or to work optimally with stand-alone lab configurations.
  • Can withstand up to 6000 psi (414 bar)
  • Chemically compatible for use with most mobile phase compositions
  • Available with analytical and nano-scale flow paths
  • Multiple control options
  • Stand alone capability
  • Included software