Chemical resistant gloves, PowerCoat® 953-04 Nitraf
Supplier: Honeywell Safety Products
112-1224EA 137 EUR
DALL2095304/9 DALL2095304/10 DALL2095304/8
Chemical resistant gloves, PowerCoat® 953-04 Nitraf
These unsupported nitrile gloves are particularly suitable for the handling of oils and greases. They also provide mechanical and chemical resistance to a large range of products (oils, detergents and many solvents).
  • Diamond finish provides good grip when handling dry and damp objects
  • Nitrile has good chemical properties and excellent mechanical characteristics (abrasion resistant)
  • Long cuff helps to protect the forearm
  • Smooth inside for easier donning
Ideal for food preparation (greasy products), the production of paints, and varnishes.
Certifications: EN 388:2016 (4101X), EN ISO 374:2016 Type A (AJKLMNOT), EN ISO 374-5:2016 (MICRO-ORGANISMS); Cat. III
Caution: Please note: Avoid contact with solvents containing ketones.