BIOV2730

BIOV2730-250GEA 282 EUR
BIOV2730-250G BIOV2730-50G
BIOV2730
Pluronic® F-127 is a non ionic detergent used for isolation of membrane proteins and also to facilitate the solubilisation of water-insoluble dyes and other materials in physiological media.

  • Appearance: white solid
  • Solubility: 50 mg/ml (H₂O)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR