Guanidinium hydrochloride 99.5-101.0%, GenAR® for biotechnology, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Guanidinium chlorideGuanidine hydrochlorideGuanidine chlorideGuanadine hydrochloride

0832-F9EA 0 EUR
0832-F9
Guanidinium hydrochloride 99.5-101.0%, GenAR® for biotechnology, Macron Fine Chemicals™
Guanidinium hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
MW: 95,53 g/mol
MDL Number: MFCD00013026
CAS Number: 50-01-1
https://ie.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (NH₂C(:NH)NH₂·HCl) 99.5 - 101.0 %
Acidity (as HCl) ≤0.01 %
Loss on Drying at 105°C ≤0.5 %
Residue after Ignition ≤0.050 %
Absorbance of a 6M Aqueous Solution - 230 nm ≤0.200
Absorbance of a 6M Aqueous Solution - 260 nm ≤0.030
Absorbance of a 6M Aqueous Solution - 275 nm ≤0.030
RNase Activity (None Detected) Passes Test
Enzyme Activity - DNase Activity Passes Test
Enzyme Activity - Protease Activity Passes Test
Nitrate (NO₃) ≤0.005 %
Sulfate (SO₄) ≤0.005 %
Arsenic and Antimony (as As) ≤0.500 ppm
Trace Impurities - Copper (Cu) ≤0.200 ppm
Cyanide (CN) ≤0.1 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Trace Impurities - Lead (Pb) ≤0.500 ppm
Particulate Matter Passes Test
MANUFACTURED EXCLUSIVELY FOR AVANTOR PERFORMANCE MATERIALS, LLC., BY
BIOSPECTRA, INC., STROUDSBURG, PA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR