Industrial gloves, Oil Grip FC, NF35F
Supplier: Honeywell Safety Products
DALLNF35F/8MEA 88.5 EUR
DALLNF35F/8M DALLNF35F/7S DALLNF35F/10XL
Industrial gloves, Oil Grip FC, NF35F
Gloves
These green polamide knitted gloves are fully Double dipped with a black nitrile coating.
  • Ergonomic seamless knit construction conforms better to the natural shape of the hand
  • Comfortable fit enables longer use and less fatigue
  • Polyamide yarn reduces linting for the most sensitive applications
  • Can be easily laundered for extended use
  • Double dipping in nitrile provides an excellent protection to liquids (oil, water)
  • Very good grip thanks to micro capillaries which draws oil and water away from the surface
Ideal for the handling of fine products in a greasy, oily or damp atmosphere. Typical industries include: Building and construction, maintenance, metal stee, mining and quarrying, offshore, ship building and transportation.
Certifications: EN 388:2016 (3131X); Cat. II