Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Immersion Oils

https://ie.vwr.com/stibo/smallweb/std.lang.all/44/12/22474412.jpg

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  Merck
Description:   Immersion media are used in conjunction with immersion objectives and are located between the surface of the specimen and the lens of the microscope. ...
Catalog Number: (630-1808)

Supplier:  VWR Collection
Description:   Immersion oil is used to increase microscope resolution.
UOM:  1 * 1 items
Catalog Number: (NIKOMXA20235)

Supplier:  NIKON UK LTD
Description:   Accessory, Immersion oil - Type A (500 ml)
UOM:  1 * 1 items
Catalog Number: (630-0340)

Supplier:  ZEISS
Description:   Immersion oil 518 F oiler 20 ml according ISO 8036-1/2.
UOM:  1 * 20 ml
Supplier:  RAL (REACTIFS RAL SA)
Description:   This immersion oil for microscopy is highly transparent and has a viscosity designed for specific and accurate use.
Supplier:  ZEISS
Description:   Chemical accessory, Bottle 500 ml immersion oil
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 6  of 6
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping