Anti-GALP Goat Polyclonal Antibody

Supplier: ProSci Inc.
PRSI45-651EA 435 EUR
PRSI45-651
Anti-GALP Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000.

GALP antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GALP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GALP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR