|
![]() |
ORDER
SPECIFICATIONS
Antigen symbol | HCAR1 |
Reactivity | Human, Monkey |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Host | Rabbit |
ELISA | Yes |
ImmunoChemistry | Yes |
Immunogen | GPR81 antibody was raised against a peptide located in the N-Terminal extracellular domain of GPR81 (Human). |
Form | Liquid |
Tested applications | Immunoaffinity Chromatography |