Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GPRC6A Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
GPRC6A has been shown to bind to L-alpha-amino acids. Recently GPRC6A has been suggested to be a calcium-sensing receptor (Pi et al. 2005).

GPRC6A antibody can be used in Western Blot starting at 0.5 - 2 μg/mL, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPRC6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG PRSI48-164
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol GPRC6A
Antigen name G Protein-coupled Receptor Class C Group 6 Member A
Reactivity Human, Gorilla
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
ImmunoChemistry Yes
Antigen synonyms bA86F4.3, GPCR
Immunogen GPRC6A antibody was raised against a peptide located in the N-Terminal extracellular domain of GPRC6A (Human).
Form Liquid
Tested applications Immunoaffinity Chromatography
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty