Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PYGB Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
PYGB antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 μg/mL, and flow cytometry starting at 1:10.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PYGB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Chicken, Drosophila, Gibbon, Gorilla, Hamster, Human, Monkey, Mouse, Orangutan, Pig, Rabbit, Rat, Sheep, Xenopus, Zebrafish




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG PRSI48-190
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol PYGB
Antigen name Phosphorylase, Glycogen; Brain
Reactivity Gibbon, Chicken, Hamster, Human, Drosophila, Zebrafish, Xenopus, Mouse, Monkey, Rat, Pig, Sheep, Gorilla, Bovine, Orangutan, Rabbit, Bat
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
ImmunoChemistry Yes
Antigen synonyms GPBB
Immunogen PYGB antibody was raised against a peptide located near the internal domain of PYGB (Human).
Form Liquid
Tested applications Immunoaffinity Chromatography
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty