Anti-GPR12 Rabbit Polyclonal Antibody

Supplier: ProSci
PRSI48-205EA 619 EUR
PRSI48-205
Anti-GPR12 Rabbit Polyclonal Antibody
Antibodies
GPR12 has been identified as a constitutively active Lysophospholipid/Lysosphingolipid receptor.

GPR12 antibody can be used in Western Blot, immunohistochemistry starting at 20 μg/mL, and flow cytometry.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Dog, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR