Sphingosine kinase (SPHK) phosphorylates sphingosine to form a bioactive lipid mediate, sphingosine 1-phosphate (S1P or SPP). S1P is a novel lipid messenger with both intracellular and extracellular functions. Intracellularly, it regulates proliferation and survival while extracellularly it is a ligand for EDG 1. Expression of SPHK1 and subsequent production of S1P promotes the G1-S transition, protects cells from apoptosis, stimulates migration and angiogenesis, and induces tumor formation in mice.
SPHK1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SPHK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human