Anti-GPR84 Rabbit Polyclonal Antibody

Supplier: ProSci
PRSI48-221EA 619 EUR
PRSI48-221
Anti-GPR84 Rabbit Polyclonal Antibody
Antibodies
G protein-coupled receptor EX33 is an Orphan-A GPCR with an unknown ligand.

GPR84 antibody can be used in ELISA, and immunohistochemistry starting at 20 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR84
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR