Anti-CCR8 Rabbit Polyclonal Antibody

Supplier: ProSci
PRSI48-226EA 619 EUR
PRSI48-226
Anti-CCR8 Rabbit Polyclonal Antibody
Antibodies
CCR8, a Chemokine Receptor, binds the CC-chemokine I-309, which is secreted by umbilical vein endothelial cells. CCR8 also binds viral monocyte inflammatory protein-I (vMIP-I), which is induced by Kaposi sarcoma-related human herpesvirus-8. CCR8 induces endothelial chemotaxis and may function as an HIV-1 co-receptor on primary human cells, contributing to HIV-1-induced thymic pathogenesis (Lee et al., 2000).

CCR8 antibody can be used in immunohistochemistry starting at 4.8 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCR8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR