|
![]() |
ORDER
SPECIFICATIONS
Antigen symbol | GPR182 |
Antigen name | G Protein-coupled Receptor 182 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Host | Rabbit |
ELISA | Yes |
ImmunoChemistry | Yes |
Antigen synonyms | 7TMR, gamrh, AM-R, ADMR, hrhAMR, G10D, AMR |
Immunogen | GPR182 antibody was raised against a peptide located in the C-Terminal domain of GPR182 (Human). |
Form | Liquid |
Tested applications | Immunoaffinity Chromatography |