Anti-DRD1 Rabbit Polyclonal Antibody

Supplier: ProSci
PRSI48-241EA 480 EUR
PRSI48-241
Anti-DRD1 Rabbit Polyclonal Antibody
DRD1, a Dopamine Receptor, mediates the effect of the neurotransmitter dopamine, an intermediate in the biosynthesis of noradrenaline and adrenaline. It is involved in the control of motor activity and cognitive memory. Downregulation of DRD1 leads to severe impairments in working memory and to locomotor hyperactivity, which are observed in diseases such as schizophrenia, Parkinson's disease, affective disorders, and attention deficit hyperactivity disorder. DRD1 enhances smooth muscle contraction and vasodilation in the heart. It also modulates sodium transport in the kidney and is involved in the pathogenesis of hypertension.

DRD1 antibody can be used in immunohistochemistry starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DRD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR