Anti-TNFRSF18 Rat Monoclonal Antibody [clone: MGIT 07]

Supplier: ProSci
PRSI36-007EA 0 EUR
PRSI36-007
Anti-TNFRSF18 Rat Monoclonal Antibody [clone: MGIT 07]
Antibodies
GITR is the receptor for TNFSF18. It is involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death. Mediates NF-kappaB activation via the TRAF2/NIK pathway.

ELISA: (direct and indirect: 1:5,000-1:10,000). Flow Cytometry: (1:1,000). Western blot (excellent): (1:2,000-1:5,000 using ECL. Suggested blocking and dilution buffer is TBST containing 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at room temperature.). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: MGIT 07
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR