Anti-HAVCR2 Rat Monoclonal Antibody [clone: TI 142F]

Supplier: ProSci
PRSI36-029EA 375 EUR
PRSI36-029
Anti-HAVCR2 Rat Monoclonal Antibody [clone: TI 142F]
Antibodies
Tim-3 is a Th1-specific cell surface protein. It is expressed selectively by differentiated CD4+Th1 and CD8+Tc1 cells, but is absent on CD4+Th2 and CD8+ Tc2 cells. The Tim gene family has an important role in the regulation of autoimmunity and allergies.

ELISA: (direct or indirect: 1:2,000-1:10,000). Flow Cytometry: (1:1,000). Optimal conditions should be determined individually for each application.

Type: Primary
Antigen: HAVCR2
Clonality: Monoclonal
Clone: TI 142F
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR