Anti-TNFSF9 Mouse Monoclonal Antibody [clone: 41B446]

Supplier: ProSci
PRSI36-064EA 0 EUR
PRSI36-064
Anti-TNFSF9 Mouse Monoclonal Antibody [clone: 41B446]
The CD137 ligand (CD137L, 4-1BBL) is a member of the tumor necrosis factor (TNF) family. CD137L is a type II, transmembrane protein found on activated macrophages, dendritic cells and mature B cells. The interaction with its receptor 4-1BB induces recruitment of TNF receptor-associated factor 1 (TRAF1) and TRAF2 and interaction with the kinase p56lck. CD137 and CD137L have been reported to be involved in tumor rejection, apoptosis, anti-viral immunity, diabetes, in T and B cell co-stimulation and modulation of the immune response.

Flow Cytometry: (1:1,000). Western Blot: (1:2,000). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: TNFSF9
Clonality: Monoclonal
Clone: 41B446
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR