Anti-IL17A Mouse Monoclonal Antibody [clone: 17-142F]

Supplier: ProSci
PRSI36-072EA 375 EUR
PRSI36-072
Anti-IL17A Mouse Monoclonal Antibody [clone: 17-142F]
Antibodies
IL-17A exhibits multiple biological activities on a variety of cells including the induction of IL-6 and IL-8 production in fibroblsts, the enhancement of surface expression of ICAM-1 in fibroblast, activation of NF-kappaB and costimulation of T cell proliferation. Induces stromal cells to produce proinflammatory and hematopoietic cytokines.

ELISA: (direct and indirect: 1:500-1:5,000). Flow Cytometry: (1:20-1:500). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 17-142F
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR