Anti-TNFRSF9 Rat Monoclonal Antibody [clone: M4173]

Supplier: ProSci
PRSI36-074EA 0 EUR
PRSI36-074
Anti-TNFRSF9 Rat Monoclonal Antibody [clone: M4173]
Antibodies
CD137 is a member of the tumor necrosis factor (TNF) receptor family. CD137 is expressed by activated T cells, dendritic cells, NK cells, granulocytes and cells of blood vessel walls at sites of inflammation. CD137 has costimulatory activity for activated T cells. Crosslinking of CD137 enhances T cell proliferation, IL-2 secretion survival and cytolytic activity. CD137 was shown to enhance immune activity to eliminate tumors in mice.

Flow Cytometry: (1:500). Western Blot: (1:2,000). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: TNFRSF9
Clonality: Monoclonal
Clone: M4173
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR