Anti-TNFRSF13C Mouse Monoclonal Antibody (Biotin) [clone: HuBR9.1]

Supplier: ProSci
PRSI36-116EA 612 EUR
PRSI36-116
Anti-TNFRSF13C Mouse Monoclonal Antibody (Biotin) [clone: HuBR9.1]
Antibodies
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.

Type: Primary
Antigen: TNFRSF13C
Clonality: Monoclonal
Clone: HuBR9.1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR