Anti-TNFRSF13C Mouse Monoclonal Antibody (ATTO 488) [clone: HuBR9.1]

Supplier: ProSci
PRSI36-117EA 514 EUR
PRSI36-117
Anti-TNFRSF13C Mouse Monoclonal Antibody (ATTO 488) [clone: HuBR9.1]
Antibodies
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.

Definition for 1 test: 1ul of labeled antibody in 200ul of Flow Cytometry buffer (see online protocol).

Type: Primary
Antigen: TNFRSF13C
Clonality: Monoclonal
Clone: HuBR9.1
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR