Anti-TNFRSF10B Mouse Monoclonal Antibody (ATTO 647N) [clone: HS201]

Supplier: ProSci
PRSI36-133EA 655 EUR
PRSI36-133
Anti-TNFRSF10B Mouse Monoclonal Antibody (ATTO 647N) [clone: HS201]
Antibodies
TRAIL-R2 is a receptor for the cytotoxic ligand TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. Promotes the activation of NF-kappaB.

Definition for 1 test: 1ul of labeled antibody in 200ul of Flow Cytometry buffer (see online protocol).

Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: HS201
Conjugation: ATTO 647N
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR