Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: HS301]

Supplier: ProSci
PRSI36-134EA 612 EUR
PRSI36-134
Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: HS301]
Antibodies
TRAIL-R3 is a receptor for the cytotoxic ligand TRAIL. It lacks a cytoplasmic death domain and hence is not capable of inducing apoptosis. Protects cells against TRAIL mediated apoptosis by competing with TRAIL-R1 and R2 for binding to the ligand.

Definition for 1 test: 1ul of labeled antibody in 200ul of Flow Cytometry buffer (see online protocol).

Type: Primary
Antigen: TNFRSF10C
Clonality: Monoclonal
Clone: HS301
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR