Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: TR2.21]

Supplier: ProSci
PRSI36-138EA 514 EUR
PRSI36-138
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: TR2.21]
Antibodies
TRAIL-R2 is a receptor for the cytotoxic ligand TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. Promotes the activation of NF-kappaB.

Immunohistochemistry: (paraffin sections) (20ug/ml). Western Blot: (1ug/ml) . Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: TR2.21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR