Anti-TNFRSF18 Rat Polyclonal Antibody

Supplier: ProSci
PRSI36-215EA 454 EUR
PRSI36-215
Anti-TNFRSF18 Rat Polyclonal Antibody
Antibodies
GITR is the receptor for TNFSF18. It is involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death. Mediates NF-kappaB activation via the TRAF2/NIK pathway.

ELISA: (direct and indirect: 1:5,000-1:10,000). Flow Cytometry: (1:1,000). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: TNFRSF18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR