Anti-TNFRSF11A Rabbit Polyclonal Antibody

Supplier: ProSci
PRSI36-219EA 312 EUR
PRSI36-219
Anti-TNFRSF11A Rabbit Polyclonal Antibody
Antibodies
RANK is the receptor for RANKL/TRANCE/OPGL. It is essential for RANKL-mediated osteoclastogenesis and is involved in the regulation of interactions between T-cells and dendritic cells.

Western Blot: (1:500-1:2,000 using ECL. Suggested blocking and dilution buffer is TBST with 0.05% Tween 20 with 5% skim milk. Suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: TNFRSF11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR