Human RTP801 peptide (14 aa near N-terminus)
Supplier: ProSci
PRSI4509PEA 100 EUR
PRSI4509P
Human RTP801 peptide (14 aa near N-terminus)
Proteins and Peptides
RTP801 peptide is used for blocking the activity of RTP801 antibody.