Human EBI3 peptide (15 aa near C-terminus)
Supplier: ProSci
PRSI4967PEA 105 EUR
PRSI4967P
Human EBI3 peptide (15 aa near C-terminus)
Proteins and Peptides
EBI3 peptide is used for blocking the activity of EBI3 antibody.