H1N1 Hemagglutinin Peptide
Supplier: ProSci
PRSI5231PEA 100 EUR
PRSI5231P
H1N1 Hemagglutinin Peptide
Proteins and Peptides
Seasonal H1N1 Hemagglutinin peptide is meant to be used as a positive control in ELISA applications to detect Seasonal Influenza A antibody.