Seasonal H1N1 hemagglutinin peptide
Supplier: ProSci
PRSI5239PEA 105 EUR
PRSI5239P
Seasonal H1N1 hemagglutinin peptide
Proteins and Peptides
Seasonal H1N1 Hemagglutinin peptide is meant to be used as a positive control in ELISA applications to detect Seasonal Influenza A antibody.