Pig H1N1 Hemagglutinin Peptide
Supplier: ProSci
PRSI5233PEA 100 EUR
PRSI5233P
Pig H1N1 Hemagglutinin Peptide
Proteins and Peptides
Swine H1N1 Hemagglutinin peptide is meant to be used as a positive control in ELISA applications to detect Swine-Origin Influenza A antibody.