Pig H1N1 Nonstructural Protein 1 Peptide
Supplier: ProSci
PRSI5359PEA 105 EUR
PRSI5359P
Pig H1N1 Nonstructural Protein 1 Peptide
Proteins and Peptides
Swine H1N1 Nonstructural Protein 1 peptide is meant to be used as a positive control in ELISA applications to detect Swine-Origin Influenza A antibody.