Human ZIP3 Peptide (16 aa in the center)
Supplier: ProSci
PRSI6085PEA 100 EUR
PRSI6085P
Human ZIP3 Peptide (16 aa in the center)
Proteins and Peptides
ZIP3 peptide is used for blocking the activity of ZIP3 antibody.