Human RILPL1 peptide (17 aa near N-terminus)
Supplier: ProSci
PRSI6495PEA 105 EUR
PRSI6495P
Human RILPL1 peptide (17 aa near N-terminus)
Proteins and Peptides
RILPL1 peptide is used for blocking the activity of RILPL1 antibody.