Recombinant IMP1 (from E. coli)
Supplier: ProSci
PRSI39-901EA 395 EUR
PRSI39-901
Recombinant IMP1 (from E. coli)
Proteins and Peptides
As Gram-negative bacterial isolates producing plasmid-mediated IMP-1 metallo-beta-lactamase usually demonstrate resistance to various broad-spectrum beta-lactams, including cephamycins and carbapenems, transmission and proliferation of these microorganisms in clinical settings could become a clinical threat in the near future.
  • This recombinant protein can be used for WB, ELISA.
20mM Tris-HCl, 150mM NaCl, 1mM EDTA, 5mM Glutathione, 0.5mM GSSH and 10% Glycerol.
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.