Recombinant Survivin (from E. coli)
Supplier: ProSci
PRSI39-904EA 395 EUR
PRSI39-904
Recombinant Survivin (from E. coli)
Proteins and Peptides
Survivin is a member of the inhibitor of apoptosis protein (IAP) family, that has been implicated in both control of cell division and inhibition of apoptosis.
  • This recombinant protein can be used for WB, ELISA.
20mM Tris-HCl, 150mM NaCl, 1mM EDTA, 5mM Glutathione, 0.5mM GSSH and 10% Glycerol.
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.