Recombinant TNF alpha (from E. coli)
Supplier: ProSci
PRSI39-907EA 395 EUR
PRSI39-907
Recombinant TNF alpha (from E. coli)
Proteins and Peptides
TNF-alpha belongs to the tumor nexrosis factor family and is believed to play roles in antitumor activity, immune moedulation, infalmmation, anorexia, cachexia, septicshock, viral replication and ematopoiesis. TNF-alpha is currently being evaluated in treatment of certain cancers and AIDS related symptoms.
  • This recombinant protein can be used for WB, ELISA.
20mM Tris-HCl, 150mM NaCl, 1mM EDTA, 5mM Glutathione, 0.5mM GSSH and 10% Glycerol.
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.