Mouse Recombinant RELM beta (from E. coli)
Supplier: ProSci
PRSI40-485EA 185 EUR
PRSI40-485
Mouse Recombinant RELM beta (from E. coli)
Proteins and Peptides
The biological functions of RELM beta, and its molecular targets, are not fully known but, it has been suggested that it plays a regulatory role during inflammation and may also act to establish links among adipose tissue, the intestine and the liver (Rajala, M. et al. J. Clin. Invest. Vol. 111, 225-230 (2003)). Interestingly the molecular structure of RELM beta is highly homologous to that of the adipose-derived cytokine Resistin and RELM beta. These proteins share a highly conserved C-terminal domain, characterized by 10 cysteine residues with a unique spacing motif of C-X11-C-X8-C-X-C-X3-C-X10-C-X-C-X-C-X9-C-C.
  • Human RELM-beta is a 19.0 kDa disulfide-linked homodimeric protein, consisting of two 89 amino acid residue chains, expressed in the epithelium of the colon and small bowel. Murine RELM-beta (Resistin-like molecule beta/FIZZ2) is an 18 kDa disulfide-linked homodimeric protein, consisting of two 83 amino acid residue chains, expressed in the epithelium of the colon and small bowel.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.