Human Recombinant CXCL4 (from E. coli)
Supplier: ProSci
PRSI40-104EA 185 EUR
PRSI40-104
Human Recombinant CXCL4 (from E. coli)
Proteins and Peptides
CXCL4 is a CXC chemokine that is expressed in megakaryocytes and stored in the alpha-granules of platelets. CXCL4 is chemotactic towards neutrophils and monocytes and has been shown to inhibit angiogenesis. Recombinant human CXCL4 is a 7.8 kDa protein containing 70 amino acid residues, including the four highly conserved residues present in CXC chemokines.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.