Human Recombinant TGF beta (from (BTI-Tn-5B1-4) Hi-5 Insect cells)
Supplier: ProSci
PRSI40-108EA 185 EUR
PRSI40-108
Human Recombinant TGF beta (from (BTI-Tn-5B1-4) Hi-5 Insect cells)
Proteins and Peptides
The three mammalian isoforms of TGF-β, TGF-β1, β2, β3, signal through the same receptor and elicit similar biological responses. They are multifunctional cytokines that regulate cell proliferation, growth, differentiation and motility as well as synthesis and deposition of the extracellular matrix. They are involved in various physiological processes including embryogenesis, tissue remodeling and wound healing. They are secreted predominantly as latent complexes which are stored at the cell surface and in the extracellular matrix. The release of biologically active TGF-β isoform from a latent complex involves proteolytic processing of the complex and /or induction of conformational changes by proteins such as thrombospondin-1. TGF-β2 has been shown to exert suppressive effects on IL-2 dependent T-cell growth, and may also have an autocrine function in enhancing tumor growth by suppressing immuno-surveillance of tumor development. Recombinant human TGF-β2 is a 25.0 kDa protein composed of two identical 112 amino acid polypeptide chains linked by a single disulfide bond.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.