Human Recombinant GMCSF (from E. coli)
Supplier: ProSci
PRSI40-281EA 185 EUR
PRSI40-281
Human Recombinant GMCSF (from E. coli)
Proteins and Peptides
GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells. It is produced in by endothelial cells, monocytes, fibroblasts and T-lymphocytes. GM-CSF inhibits neutrophil migration and enhances the functional activity of the mature end-cells. The human and murine molecules are species-specific and exhibit no cross-species reactivity. Recombinant human GM-CSF is a 14.6 kDa globular protein consisting of 128 amino acids containing two intramolecular disulfide bonds and two potential N-linked glycosylation sites.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.