Human Recombinant Mat2A (from E. coli)
Supplier: ProSci
PRSIXW-RP3143EA 530 EUR
PRSIXW-RP3143
Human Recombinant Mat2A (from E. coli)
Proteins and Peptides
Methionine adenosyltransferase (EC 2.5.1.6) catalyzes the biosynthesis of S-adenosylmethionine (AdoMet) from methionine and ATP. AdoMet is the major methyl donor for many of the transmethylation reactions in the body. The MAT2A promoter is highly GC rich, especially in the first 300 bp. The promoter contains several potential SP1-, v-myb-, and GATA-binding sites, as well as potential binding sites for C/EBP, HSF2, c-myb, and STATx. It can efficiently drive expression from a reporter gene in both Jurkat and 293 cells.
  • This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.