Human Recombinant G-CSF (from E. coli)
Supplier: ProSci
PRSI40-287EA 185 EUR
PRSI40-287
Human Recombinant G-CSF (from E. coli)
Proteins and Peptides
G-CSF is a hematopoietic growth factor that stimulates the development of committed progenitor cells to neutrophils and enhances the functional activities of the mature end-cell. It is produced in response to specific stimulation by a variety of cells including macrophages, fibroblasts, endothelial cells and bone marrow stroma. G-CSF is being used clinically to facilitate hematopoietic recovery after bone marrow transplantation. Human and murine G-CSF are cross-species reactive. Recombinant human G-CSF is an 18.7 kDa protein consisting of 174 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.