Human Recombinant Leptin (from E. coli)
Supplier: ProSci
PRSI40-289EA 185 EUR
PRSI40-289
Human Recombinant Leptin (from E. coli)
Proteins and Peptides
Encoded by the ob (obese) gene, Leptin is an adipose-derived cytokine that suppresses appetite and increases thermogenesis. Leptin exerts its anorectic effect via signaling through a hypothalamic receptor termed OB-R. Leptin has been shown to reduce body weight, food consumption, and plasma glucose levels in various in vivo models. Recombinant human Leptin is a 16.0 kDa protein containing 147 amino acid residues. Recombinant murine Leptin is a 16.2 kDa protein containing 147 amino acid residues. Recombinant Rat Leptin is a 16.2 kDa protein containing 147 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.