Human Recombinant Trefoil Factor 2 (from E. coli)
Supplier: ProSci
PRSI40-291EA 185 EUR
PRSI40-291
Human Recombinant Trefoil Factor 2 (from E. coli)
Proteins and Peptides
The Trefoil Factor peptides (TFF1, TFF2 and TFF3) are expressed in the gastrointestinal tract, and appear to play an important role in intestinal mucosal defense and repair. TFF2 has been shown to inhibit gastrointestinal motility and gastric acid secretion. Recent data suggests a potential role for TFF2 in acute and chronic asthma (Nikolaidis, N.M. et al. Am. Journal Respir. Cell Mol. Biol. (2003) 4: 458-464). Recombinant human TFF2 is a 12.0 kDa polypeptide of 106 amino acid residues, which includes a 40-amino acid trefoil motif containing three conserved intramolecular disulfide bonds.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.