Human Recombinant TNF Receptor II (soluble) (from E. coli)
Supplier: ProSci
PRSI40-304EA 185 EUR
PRSI40-304
Human Recombinant TNF Receptor II (soluble) (from E. coli)
Proteins and Peptides
TNFRII is a member of the TNFR family of transmembrane proteins, and is expressed in immune cells and certain endothelial cells. It is a high affinity receptor for TNF-α but manifests a lower affinity to TNF-β. Signaling through this receptor regulates various biological processes including cell proliferation, differentiation, apoptosis, lipid metabolism, coagulation, and neurotransmission. Soluble TNFRII is capable of inhibiting TNF-α induced activities by acting as a decoy receptor. Recombinant human sTNFRII is an 18.9 kDa protein (174 amino acid residues) comprising the cysteine rich ligand binding portion of the extracellular domain of the TNFRII protein.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.