Human Recombinant Hvem-Fc (from Hi-5 Insect cells)
Supplier: ProSci
PRSI40-316EA 285 EUR
PRSI40-316
Human Recombinant Hvem-Fc (from Hi-5 Insect cells)
HVEM belongs to the TNF Receptor superfamily of transmembrane proteins and plays a role in the activation of T-cells and other lymphocytes. It is expressed in various cells and tissues including spleen, thymus, lung, macrophages, and T-cells. HVEM activation induces a signaling cascade which results in induction of transcription factors NF-kappaB and AP-1. LIGHT (TNFSF14) and TNF-beta (TNFSF1) function as the ligands for HVEM, which can also bind specifically to herpes simplex virus glycoprotein D. Soluble HVEM can act as a "receptor decoy” resulting in inhibition of the activity of the HVEM ligands, LIGHT and TNF-beta. Recombinant human HVEM-Fc Chimera is a 376 amino acid fusion protein containing an N-terminal domain corresponding to the extracellular region of HVEM and a C-terminal domain corresponding to residues 102 to 330 of human IgG1.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.